Learn More
PeproTech Epo Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
£177.00 - £1468.00
Specifications
| Antigen | Epo |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17855373
|
PeproTech
500-P318-50UG |
50 μg |
£177.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17865793
|
PeproTech
500-P318-100UG |
100 μg |
£228.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17845333
|
PeproTech
500-P318-1MG |
1 mg |
£1468.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: APPRLICDSR VLERYLLEAK EAENITTGCA EHCSLNENIT VPDTKVNFYA WKRMEVGQQA VEVWQGLALL SEAVLRGQAL LVNSSQPWEP LQLHVDKAVS GLRSLTTLLR ALGAQKEAIS PPDAASAAPL RTITADTFRK LFRVYSNFLR GKLKLYTGEA CRTGDR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human EPO. Anti-Human EPO-specific antibody was purified by affinity chromatography employing an immobilized Human EPO matrix. Sandwich ELISA: To detect Human EPO by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human EPO (500-P318Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human EPO. Western Blot: To detect Human EPO by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human EPO is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.Specifications
| Epo | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P01588 | |
| 2056 | |
| CHO cells-derived Recombinant Human EPO. | |
| Antigen affinity chromatography | |
| EPO |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2 | |
| EPO | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.