missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPN1 Mouse anti-Human, Clone:6F7F9, Abnova™
Mouse monoclonal antibody raised against recombinant human EPN1.
Brand: Abnova MAB16651.100ug
This item is not returnable.
View return policy
Description
EPN1 is an endocytic accessory protein that interacts with EPS15 (MIM 600051), the alpha subunit of the clathrin adaptor AP2 (AP2A1; MIM 601026), and clathrin (see MIM 118960), as well as with other accessory proteins for the endocytosis of clathrin-coated vesicles.[supplied by OMIM]Specifications
| EPN1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| EPN1 | |
| Recombinant protein corresponding to amino acid 106-254 of human EPN1 from E. coli. | |
| 29924 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
| ELISA, Flow Cytometry, Western Blot | |
| 6F7F9 | |
| epsin 1 | |
| In PBS (0.05% sodium azide) | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction