missing translation for 'onlineSavingsMsg'
Learn More

EPN, TRC

Product Code. 30380830
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380830 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380830 Supplier Toronto Research Chemicals Supplier No. TRCE58890025MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EPN
CAS 2104-64-5
Molecular Formula C14 H14 N O4 P S
Quantity 25 mg
Synonym Phosphonothioic acid, phenyl-, O-ethyl O-(4-nitrophenyl) ester (9CI), Phosphonothioic acid, phenyl-, O-ethyl O-(p-nitrophenyl) ester (8CI), Phosphonothioic acid, phenyl-, ethyl p-nitrophenyl ester (7CI), EPN, EPN 300, NSC 404840, NSC 8943, O-ETHYL O-(P-NITROPHENYL) PHENYLPHOSPHONOTHIOATE, O-Ethyl O-(4-nitrophenyl) benzenethionophosphonate, O-Ethyl O-(4-nitrophenyl) phenylphosphonothioate, O-Ethyl O-(p-nitrophenyl) phenylphosphonothioate, O-Ethyl O-p-nitrophenyl benzenethiophosphonate, Santox
InChI Formula InChI=1S/C14H14NO4PS/c1-2-18-20(21,14-6-4-3-5-7-14)19-13-10-8-12(9-11-13)15(16)17/h3-11H,2H2,1H3
SMILES CCOP(=S)(Oc1ccc(cc1)[N+](=O)[O-])c2ccccc2
IUPAC Name ethoxy-(4-nitrophenoxy)-phenyl-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 323.3
Formula Weight 323.0381
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.