missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPM2AIP1 Antibody (OTI2A2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71497DL594
This item is not returnable.
View return policy
Description
EPM2AIP1 Monoclonal antibody specifically detects EPM2AIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| EPM2AIP1 | |
| Monoclonal | |
| DyLight 594 | |
| EPM2A (laforin) interacting protein 1, EPM2A interacting protein 1, EPM2A-interacting protein 1, KIAA0766FLJ11207, Laforin-interacting protein | |
| Full length recombinant protein of human EPM2AIP1 (NP_055620) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2A2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 9852 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction