missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPM2AIP1 Antibody (OTI2A2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71497CL1
This item is not returnable.
View return policy
Description
EPM2AIP1 Monoclonal antibody specifically detects EPM2AIP1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| EPM2AIP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| EPM2A (laforin) interacting protein 1, EPM2A interacting protein 1, EPM2A-interacting protein 1, KIAA0766FLJ11207, Laforin-interacting protein | |
| Full length human recombinant protein of human EPM2AIP1(NP_055620) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI2A2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 9852 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction