missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPM2AIP1 Antibody (OTI2A2), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71497AF647
This item is not returnable.
View return policy
Description
EPM2AIP1 Monoclonal specifically detects EPM2AIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| EPM2AIP1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EPM2AIP1 | |
| Full length recombinant protein of human EPM2AIP1 (NP_055620) produced in HEK293T cells. | |
| 0.1 mL | |
| 9852.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2A2 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| EPM2A (laforin) interacting protein 1, EPM2A interacting protein 1, EPM2A-interacting protein 1, KIAA0766FLJ11207, Laforin-interacting protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction