missing translation for 'onlineSavingsMsg'
Learn More

Eplivanserin (mixture), MedChemExpress

Product Code. p-200100824
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494158 10 mg 10mg
16466867 5 mg 5mg
16445126 100 mg 100mg
16400517 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494158 Supplier Medchem Express Supplier No. HY10792A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eplivanserin mixture (SR-46349 mixture) is a selective serotonin reuptake inhibitor and a 5-HT2A receptor antagonist, extracted from patent WO 2005/002578 A1.

Eplivanserin mixture (SR-46349 mixture) is a selective serotonin reuptake inhibitor and a 5-HT2A receptor antagonist, extracted from patent WO 2005/002578 A1.

Specifications

Chemical Name or Material Eplivanserin (mixture)
CAS 130581-13-4
Color Pink
Purity Grade Notes Research
Molecular Formula C19H21FN2O2
Quantity 10 mg
Synonym SR-46349 (mixture)
Solubility Information DMSO : 125 mg/mL (380.66 mM; Need ultrasonic)
SMILES OC1=CC=C(/C=C/C(C2=CC=CC=C2F)=N/OCCN(C)C)C=C1
Molecular Weight (g/mol) 328.38
Formula Weight 328.38
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.