missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Eplivanserin hemifumarate
GREENER_CHOICE

Product Code. p-7110782
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18738218

Brand: Tocris Bioscience™ 4958/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective 5-HT2A antagonist

Eplivanserin hemifumarate is a potent and selective 5-HT2A antagonist (IC50 values are 5.8 and 120 and >100 nM for 5-HT2A, 5-HT2B and 5-HT2C, respectively). Attenuates cocaine-induced hyperactivity. Increases dopamine (DA) release in rat medial prefrontal cortex (mPFC); potentiates haloperidol-induced DA release in the mPFC and nucleus accumbens.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1Z,2E)-1-(2-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one O-[2-(dimethylamino)ethyl]oxime hemifumarate
CAS 130580-02-8
Synonym SR-46349 hemifumarate, SR-46349B
Quantity 10 mg
Target 5-HT2A Receptor Antagonists
Molecular Formula C19H21FN2O2.0.5C4H4O4
Formula Weight 386.42
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.