missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPLIN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554904
This item is not returnable.
View return policy
Description
Immunogen sequence: ISENSYSLDD LEIGPGQLSS STFDSEKNES RRNLELPRLS ETSIKDRMAK YQAAVSKQSS STNYTNELKA SGGEIKIHKM EQKENVPPGP EVCITHQEGE KISANENSLA VRSTPAEDDS RDS Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
Specifications
| EPLIN | |
| Polyclonal | |
| Unconjugated | |
| LIMA1 | |
| 1110021C24Rik; D15Ertd366e; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; eplin; LIM domain and actin binding 1; LIM domain and actin-binding protein 1; LIMA1; mEPLIN; PP624; SREBP3; sterol regulatory element binding protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51474 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UHB6 | |
| LIMA1 | |
| Recombinant protein corresponding to Human EPLIN. Recombinant protein control fragment (Product #RP-91594). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction