Learn More
EPLIN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562659
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RPHKDLWASK NENEEILERP AQLANARETP HSPGVEDAPI AKVGVLAASM EAKASSQQEK EDKPAETKKL RIAWPPPTEL GSSGSALEEG I Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 82%.
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.Specifications
EPLIN | |
Polyclonal | |
Unconjugated | |
LIMA1 | |
1110021C24Rik; D15Ertd366e; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; eplin; LIM domain and actin binding 1; LIM domain and actin-binding protein 1; LIMA1; mEPLIN; PP624; SREBP3; sterol regulatory element binding protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51474 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UHB6 | |
LIMA1 | |
Recombinant protein corresponding to Human EPLIN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only