missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPLIN Monoclonal Antibody (OTI9F3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527021
This item is not returnable.
View return policy
Description
EPLIN Monoclonal Antibody for Western Blot, IHC (P)
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
Specifications
| EPLIN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UHB6 | |
| LIMA1 | |
| Human recombinant protein fragment corresponding to amino acids 1-275 of LIMA1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9F3 | |
| Unconjugated | |
| LIMA1 | |
| 1110021C24Rik; D15Ertd366e; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; eplin; LIM domain and actin binding 1; LIM domain and actin-binding protein 1; LIMA1; mEPLIN; PP624; SREBP3; sterol regulatory element binding protein 3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51474 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction