missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Eplerenone
GREENER_CHOICE

Product Code. p-4783944
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18054612

Brand: Tocris Bioscience™ 2397/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective mineralocorticoid receptor antagonist

Selective mineralocorticoid (aldosterone) receptor antagonist (IC50 = 360 nM). Displays > 27-fold selectivity over androgen, progesterone and estrogen receptors (IC50 > 10 μM). Orally active antihypertensive in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107724-20-9
Molecular Formula C24H30O6

Specifications

Quantity 10 mg
Formula Weight 414.5
Purity >99%
Molecular Weight (g/mol) 417.516
Chemical Name or Material Eplerenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.