Learn More
Volt/Ohm Meter for Epithelial Tissue Culture TEER 240V 50Hz With STX2 electrode
Brand:  WPI World Precision Instrument EVOM2
Encompass_Preferred
Product Code. 15169112
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The EVOM was the first instrument designed specifically to perform routine Trans Epithelial Electrical Resistance (TEER) measurement in tissue culture research. EVOM2 is the next generation, redesigned for ease of use. The EVOM2 not only qualitatively measures cell monolayer health, but also quantitatively measures cellular confluence. The unique electronic circuit of the EVOM2 and the included STX2 electrode detect the confluence of the cellular monolayer. When combined with WPI’s Endohm chamber, the EVOM2 can also be used to perform more accurate quantitative measurements or lower resistance measurements like trans endothelial electrical resistance measurements.