missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epithelial Stromal Interaction 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69247
This item is not returnable.
View return policy
Description
Epithelial Stromal Interaction 1 Polyclonal specifically detects Epithelial Stromal Interaction 1 in Human samples. It is validated for Western Blot.
Specifications
| Epithelial Stromal Interaction 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BRESI1, EC 3.1.4.16, EC 3.6.3.6, epithelial stromal interaction 1 (breast), epithelial-stromal interaction protein 1, MGC29634 | |
| Rabbit | |
| 47 kDa | |
| 100 μL | |
| Primary | |
| Canine: 86%; Bovine: 86%; . | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96J88 | |
| EPSTI1 | |
| Synthetic peptides corresponding to EPSTI1(epithelial stromal interaction 1 (breast)) The peptide sequence was selected from the N terminal of Epithelial Stromal Interaction 1. Peptide sequence RRLGGSQSETEVRQKQQLQLMQSKYKQKLKREESVRIKKEAEEAELQKMK. | |
| Affinity purified | |
| RUO | |
| 94240 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction