missing translation for 'onlineSavingsMsg'
Learn More

epithelial Sodium Channel beta Antibody, Novus Biologicals™

Product Code. 18367492 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367492 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367492 Supplier Novus Biologicals Supplier No. NBP318257

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

epithelial Sodium Channel beta Polyclonal antibody specifically detects epithelial Sodium Channel beta in Human, Mouse, Rat, Hamster, Xenopus samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen epithelial Sodium Channel beta
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100
Formulation PBS, 50% glycerol
Gene Alias amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta
Host Species Rabbit
Immunogen Produced against the C-terminal tail (amino acids 617-638) of rat beta ENaC (antibody designation 3755-2)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6338
Target Species Human, Mouse, Rat, Hamster, Xenopus species
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.