missing translation for 'onlineSavingsMsg'
Learn More

epithelial Sodium Channel beta Antibody (7B8), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-59380-25ug

 View more versions of this product

Product Code. 18673648

  • £168.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

epithelial Sodium Channel beta Monoclonal specifically detects epithelial Sodium Channel beta in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

epithelial Sodium Channel beta
Monoclonal
Unconjugated
PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide
SCNN1B
Synthetic peptide from the C-terminal of Rat ENaC beta (aa. 617-638)
25 μg
Phospho Specific
6338
Store at -20°C.
IgG1
Western Blot, Immunohistochemistry
7B8
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta
Mouse
Protein G purified
RUO
Primary
Detects 87kDa.
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only