missing translation for 'onlineSavingsMsg'
Learn More
Learn More
epithelial Sodium Channel beta Antibody (16E4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59381-25ug
This item is not returnable.
View return policy
Description
epithelial Sodium Channel beta Monoclonal specifically detects epithelial Sodium Channel beta in Mouse samples. It is validated for Western Blot.
Specifications
| epithelial Sodium Channel beta | |
| 160000 | |
| Western Blot 1:100 - 1:2000 | |
| amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects 87kDa. | |
| Purified |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| SCNN1B | |
| Synthetic peptide from the C-terminal of Rat ENaC beta (aa. 617-638) | |
| 25 μg | |
| Phospho Specific | |
| 6338 | |
| Store at -20°C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction