missing translation for 'onlineSavingsMsg'
Learn More
Learn More
epithelial Sodium Channel alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16353
This item is not returnable.
View return policy
Description
epithelial Sodium Channel alpha Polyclonal antibody specifically detects epithelial Sodium Channel alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| epithelial Sodium Channel alpha | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| alpha ENaC-2, alpha-ENaC, Alpha-NaCH, amiloride-sensitive epithelial sodium channel alpha subunit, amiloride-sensitive sodium channel subunit alpha, amiloride-sensitive sodium channel subunit alpha 2, BESC2, ENaCA, ENaCalpha, Epithelial Na(+) channel subunit alpha, FLJ21883, nasal epithelial sodium channel alpha subunit, Nonvoltage-gated sodium channel 1 subunit alpha, SCNEA, SCNN1, sodium channel, nonvoltage-gated 1 alpha | |
| Rabbit | |
| 76 kDa | |
| 0.1 mL | |
| Primary | |
| Expected to cross react based on sequence identity: Mouse (84%), Pig (85%), Rat (85%), Bovine (85%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P37088 | |
| SCNN1A | |
| Recombinant protein encompassing a sequence within the center region of human ENaC Alpha. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 6337 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction