missing translation for 'onlineSavingsMsg'
Learn More

epithelial Sodium Channel alpha Antibody, Novus Biologicals™

Product Code. 18433301 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433301 0.1 mL 0.1mL
18477980 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18433301 Supplier Novus Biologicals Supplier No. NBP184846

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

epithelial Sodium Channel alpha Polyclonal antibody specifically detects epithelial Sodium Channel alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen epithelial Sodium Channel alpha
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias alpha ENaC-2, alpha-ENaC, Alpha-NaCH, amiloride-sensitive epithelial sodium channel alpha subunit, amiloride-sensitive sodium channel subunit alpha, amiloride-sensitive sodium channel subunit alpha 2, BESC2, ENaCA, ENaCalpha, Epithelial Na(+) channel subunit alpha, FLJ21883, nasal epithelial sodium channel alpha subunit, Nonvoltage-gated sodium channel 1 subunit alpha, SCNEA, SCNN1, sodium channel, nonvoltage-gated 1 alpha
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: RYPEIKEELEELDRITEQTLFDLYKYSSFTTLVAGSRSRRDLRGTLPHPLQRLRVPPPPHGARRARSVASSLRDNNPQVDWKDWKIGFQL
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6337
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.