missing translation for 'onlineSavingsMsg'
Learn More

epithelial Sodium Channel alpha Antibody (2G4), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18427384 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427384 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427384 Supplier Novus Biologicals Supplier No. NBP259382JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

epithelial Sodium Channel alpha Monoclonal antibody specifically detects epithelial Sodium Channel alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen epithelial Sodium Channel alpha
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 2G4
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias alpha ENaC-2, alpha-ENaC, Alpha-NaCH, amiloride-sensitive epithelial sodium channel alpha subunit, amiloride-sensitive sodium channel subunit alpha, amiloride-sensitive sodium channel subunit alpha 2, BESC2, ENaCA, ENaCalpha, Epithelial Na(+) channel subunit alpha, FLJ21883, nasal epithelial sodium channel alpha subunit, Nonvoltage-gated sodium channel 1 subunit alpha, SCNEA, SCNN1, sodium channel, nonvoltage-gated 1 alpha
Host Species Mouse
Immunogen Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6337
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.