missing translation for 'onlineSavingsMsg'
Learn More
Learn More
epithelial Sodium Channel alpha Antibody (2G4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59382AF700
This item is not returnable.
View return policy
Description
epithelial Sodium Channel alpha Monoclonal specifically detects epithelial Sodium Channel alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
Specifications
| epithelial Sodium Channel beta | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SCNN1B | |
| Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68) | |
| 100 μL | |
| Primary | |
| Detects 85kDa. | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 2G4 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta | |
| Mouse | |
| Protein G purified | |
| Phospho Specific | |
| 6338 | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction