missing translation for 'onlineSavingsMsg'
Learn More

Epitestosterone Sulfate Triethylamine Salt, TRC

Product Code. 30383880
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383880 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383880 Supplier Toronto Research Chemicals Supplier No. TRCE588860100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Epitestosterone Sulfate Triethylamine Salt
CAS 182296-42-0
Molecular Formula C25H43NO5S
Quantity 100 mg
Synonym Androst-4-en-3-one, 17-(sulfooxy)-, (17α)-, compd. with N,N-diethylethanamine (1:1), Epitestosterone 17-Sulfate Triethylammonium Salt
InChI Formula InChI=1S/C19H28O5S.C6H15N/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(24-25(21,22)23)19(15,2)10-8-16(14)18;1-4-7(5-2)6-3/h11,14-17H,3-10H2,1-2H3,(H,21,22,23);4-6H2,1-3H3/t14-,15-,16-,17+,18-,19-;/m0./s1
SMILES CCN(CC)CC.C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@H]2OS(=O)(=O)O
Molecular Weight (g/mol) 469.68
Formula Weight 469.2862
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.