missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EpiSerf Medium
GREENER_CHOICE

Product Code. 11590356
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11590356

Brand: Thermo Scientific™ 10732022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Gibco™ EpiSerf Medium is a serum-free medium for the culture of a wide variety of adherent cells, including primary epithelial cells, MDBK, MDCK, VERO, and CHO.

The growth and morphology is comparable to the same cells grown in a classic medium supplemented with Fetal Bovine Serum (FBS). Gibco™ Episerf Medium is a confidential formulation containing L-glutamine and 2 g/L total protein.

Product Use.

For Research Use Only. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System.

Gibco™ EpiSerf Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line CHO, VERO, MDCK, MDBK
Cell Type Epithelial Cells
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Quantity 500 mL
Endotoxin Level ≤10 EU/mL
Content And Storage Storage conditions:2°C to 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Culture Type Mammalian Cell Culture
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.