missing translation for 'onlineSavingsMsg'
Learn More

Epirubicin Hydrochloride, Mikromol™

Product Code. 18071100
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18071100 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18071100 Supplier Mikromol Supplier No. MM0307.00
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Epirubicin Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Epirubicin Hydrochloride
CAS 56390-09-1
Analyte or Component Names Epirubicin Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C27 H29 N O11 . Cl H
Certifications/Compliance ISO 9001
Synonym 4'-Epidoxorubicin hydrochloride, 4'-epi-Adriamycin hydrochloride, Ellence, Epidoxorubicin hydrochloride, Epirubicin hydrochloride, Farmorubicin, Pharmorubicin, (8S,10S)-10-[(3-Amino-2,3,6-trideoxy-alpha-l-arabino-hexopyranosyl)oxy]-6,8,11-trihydroxy-8-(hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride, 5,12-Naphthacenedione, 10-[(3-amino-2,3,6-trideoxy-alpha-L-arabino-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-, hydrochloride (1:1), (8S,10S)-, 5,12-Naphthacenedione, 10-[(3-amino-2,3,6-trideoxy-alpha-L-arabino-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-(hydroxyacetyl)-1-methoxy-, hydrochloride, (8S,10S)- (9CI), 5,12-Naphthacenedione, 10-[(3-amino-2,3,6-trideoxy-alpha-L-arabino-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-(hydroxyacetyl)-1-methoxy-, hydrochloride, (8S-cis)-
InChI Formula InChI=1S/C27H29NO11.ClH/c1-10-22(31)13(28)6-17(38-10)39-15-8-27(36,16(30)9-29)7-12-19(15)26(35)21-20(24(12)33)23(32)11-4-3-5-14(37-2)18(11)25(21)34;/h3-5,10,13,15,17,22,29,31,33,35-36H,6-9,28H2,1-2H3;1H/t10-,13-,15-,17-,22-,27-;/m0./s1
SMILES Cl.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(=O)CO
IUPAC Name (7S,9S)-7-[(2R,4S,5R,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
Molecular Weight (g/mol) 579.98
Quantity 100 mg
Formula Weight 579.1507 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.