missing translation for 'onlineSavingsMsg'
Learn More

Epirubicin hydrochloride, MedChemExpress

Product Code. 16498447
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498447 5 mg 5mg
16455778 10 mg 10mg
16431937 50 mg 50mg
16406256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498447 Supplier Medchem Express Supplier No. HY13624A
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Epirubicin hydrochloride (4'-Epidoxorubicin hydrochloride), a semisynthetic L-arabino derivative of doxorubicin, has an antineoplastic agent by inhibiting Topoisomerase. Epirubicin hydrochloride inhibits DNA and RNA synthesis. Epirubicin hydrochloride is a Forkhead box protein p3 (Foxp3) inhibitor and inhibits regulatory T cell activity.

Epirubicin hydrochloride (4'-Epidoxorubicin hydrochloride), a semisynthetic L-arabino derivative of doxorubicin, has an antineoplastic agent by inhibiting Topoisomerase. Epirubicin hydrochloride inhibits DNA and RNA synthesis. Epirubicin hydrochloride is a Forkhead box protein p3 (Foxp3) inhibitor and inhibits regulatory T cell activity.

Specifications

Chemical Name or Material Epirubicin hydrochloride
CAS 56390-09-1
Color Orange
Purity Grade Notes Research
Molecular Formula C27H30ClNO11
Quantity 5 mg
Synonym 4'-Epidoxorubicin hydrochloride
Solubility Information H2O : 50 mg/mL (86.21 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (43.10 mM; Need ultrasonic)
SMILES O=C(C1=C2C(O)=C3[C@@H](O[C@@]4([H])C[C@H](N)[C@@H](O)[C@H](C)O4)C[C@@](C(CO)=O)(O)CC3=C1O)C5=CC=CC(OC)=C5C2=O.Cl
Molecular Weight (g/mol) 579.98
Formula Weight 579.98
Percent Purity 95.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.