missing translation for 'onlineSavingsMsg'
Learn More

Epiregulin Antibody (6H12), DyLight 594, Novus Biologicals™

Product Code. 18402985 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402985 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402985 Supplier Novus Biologicals Supplier No. NBP221992DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Epiregulin Monoclonal antibody specifically detects Epiregulin in Mouse samples. It is validated for ELISA, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epiregulin
Applications ELISA, Neutralization
Classification Monoclonal
Clone 6H12
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias epiregulin, ER, proepiregulin
Host Species Rat
Immunogen Mouse recombinant Epiregulin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 2069
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.