missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epiregulin Antibody (183629) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB14251JF585
This item is not returnable.
View return policy
Description
Epiregulin Monoclonal antibody specifically detects Epiregulin in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Epiregulin | |
| Monoclonal | |
| Janelia Fluor 585 | |
| epiregulin, ER, proepiregulin | |
| E. coli-derived recombinant human Epiregulin, Val63-Leu108, Accession # O14944 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 183629 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins | |
| 2069 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction