missing translation for 'onlineSavingsMsg'
Learn More

Epiregulin Antibody (183625) [DyLight 650], Novus Biologicals™

Product Code. 30737922 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737922 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737922 Supplier Novus Biologicals Supplier No. FAB1425W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Epiregulin Monoclonal antibody specifically detects Epiregulin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epiregulin
Applications Western Blot
Classification Monoclonal
Clone 183625
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias epiregulin, ER, proepiregulin
Host Species Mouse
Immunogen E. coli-derived recombinant human Epiregulin, Val63-Leu108, Accession # O14944
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 2069
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.