missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Epinephrine (+)-bitartrate salt, 98+%

Product Code. 11996881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11996881 1 g Glass bottle 1g
11926891 5 g Glass bottle 5g
11936891 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11996881 Supplier Thermo Scientific Chemicals Supplier No. 430140010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

(-)-Epinephrine (+)-bitartrate salt, CAS # 51-42-3, also known as adrenaline, is a monoamine and catecholamine hormone.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • (-)-Epinephrine (+)-bitartrate salt, is a hormone
  • It is a monoamine and catecholamine

Applications

  • (-)-Epinephrine (+)-bitartrate salt is α-adrenergic and β-adrenergic receptor agonist
  • It can be used in adipose region to minimize blood loss
  • It is potentially a platelet agonist in preparation of optimal platelet aggregometry 96-well plates
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-42-3
Molecular Formula C13H19NO9
Molecular Weight (g/mol) 333.29
MDL Number MFCD00035077
InChI Key YLXIPWWIOISBDD-NDAAPVSOSA-N
Synonym L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate
PubChem CID 124080922
SMILES O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 155.0°C
Color White to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 1 g
Merck Index 15,3674
Specific Rotation -20° to -24° (20°C, 589nm) (c=4, 1M HCl)
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Formula Weight 333.29
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material (-)-Epinephrine (+)-bitartrate salt
Show More Show Less
compliance-icons
Product Identifier
  • (-)-Epinephrine (+)-bitartrate salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.