missing translation for 'onlineSavingsMsg'
Learn More

Epinastine, MedChemExpress

Product Code. 16472186
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472186 500 mg 500mg
16418406 100 mg 100mg
16414437 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472186 Supplier Medchem Express Supplier No. HYB0640
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Epinastine (WAL801) is an antihistamine and mast cell stabilizer. Epinastine is a potent, selective and orally-active histamine H1 receptor antagonist. Epinastine also inhibits IL-8 release and has an antiallergic action.

Epinastine (WAL801) is an antihistamine and mast cell stabilizer. Epinastine is a potent, selective and orally-active histamine H1 receptor antagonist. Epinastine also inhibits IL-8 release and has an antiallergic action.

Specifications

Chemical Name or Material Epinastine
CAS 80012-43-7
Color White
Purity Grade Notes Research
Molecular Formula C16H15N3
Quantity 500 mg
Synonym WAL801
Solubility Information DMSO : ≥ 50 mg/mL (200.55 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES NC1=NCC2N1C3=CC=CC=C3CC4=CC=CC=C24
Molecular Weight (g/mol) 249.31
Formula Weight 249.31
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.