missing translation for 'onlineSavingsMsg'
Learn More

Epimedin B, MedChemExpress

Product Code. 16498918
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498918 10 mg 10mg
16464797 50 mg 50mg
16411368 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498918 Supplier Medchem Express Supplier No. HYN0259
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Epimedin B, a component extracted from Epimedii Folium, is reported to have antiosteoporotic activity.

Epimedin B, a component extracted from Epimedii Folium, is reported to have antiosteoporotic activity.

Specifications

Chemical Name or Material Epimedin B
CAS 110623-73-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C38H48O19
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (123.64 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)[C@H](OC2=CC(O)=C(C(C(O[C@@H]3O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O[C@@]4([H])[C@H](O)[C@@H](O)[C@H](O)CO4)=C(C5=CC=C(OC)C=C5)O6)=O)C6=C2C/C=C(C)/C)O1
Molecular Weight (g/mol) 808.78
Formula Weight 808.78
Percent Purity 99.61%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.