missing translation for 'onlineSavingsMsg'
Learn More

Epimedin A1, MedChemExpress

Product Code. 16488918
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16488918

Brand: Medchem Express HYN0258

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Epimedin A1 is a flavonoid extracted from Herba Epimedii which is one of commonly used Chinese medicines.

Epimedin A1 is a flavonoid extracted from Herba Epimedii which is one of commonly used Chinese medicines.

Specifications

Chemical Name or Material Epimedin A1
CAS 140147-77-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C39H50O20
Quantity 10 mg
Synonym Hexandraside F
Solubility Information DMSO : 100 mg/mL (119.22 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](O)[C@@H](O)[C@H](OC2=CC(O)=C(C(C(O[C@H]3[C@H](O)[C@H](O[C@@]4([H])[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O4)[C@@H](O)[C@H](C)O3)=C(C5=CC=C(OC)C=C5)O6)=O)C6=C2C/C=C(C)/C)O[C@@H]1CO
Molecular Weight (g/mol) 838.8
Formula Weight 838.8
Percent Purity 99.83%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.