missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epigen Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564953
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: ALTEEAAVTVT PPITAQQGNW TVNKTEADNI EGPIALKFSH LCLEDHNSYC INGACAFHHE LEKAICRCFT GYTGERCEHL TLTSYAVDSY E Highest antigen sequence identity to the following orthologs - mouse 83%, rat 80%.
Epigen is an EGF-related polypeptide growth factor that signals through the ErbB receptor-1. Promotes the growth of epithelial cells. May stimulate the phosphorylation of EGFR and mitogen-activated protein kinases. It is produced in several tissues, including the testis, liver, and heart, as well as in certain tumor cells. Epigen is mitogenic for fibroblasts and epithelial cells. Human Epigen is initially synthesized as a glycosylated 14.7 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a mature soluble sequence.
Spécification
| Epigen | |
| Polyclonal | |
| Unconjugated | |
| EPGN | |
| 2310069M11Rik; ALGV3072; EPG; EPGN; epigen; epithelial mitogen; epithelial mitogen homolog; PRO9904; RGD1560084; UNQ3072/PRO9904 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 255324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UW88 | |
| EPGN | |
| Recombinant Human Epigen. Recombinant protein control fragment (Product #RP-90531). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu