missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Epigallocatechin Gallate, TRC
Specifications
Specifications
| Chemical Name or Material | (-)-Epigallocatechin gallate |
| CAS | 989-51-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H18 O11 |
| Quantity | 10 g |
| Synonym | Benzoic acid, 3,4,5-trihydroxy-, (2R,3R)-3,4-dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-2H-1-benzopyran-3-yl ester, Benzoic acid, 3,4,5-trihydroxy-, 3,4-dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-2H-1-benzopyran-3-yl ester, (2R-cis)-, Epigallocatechol, 3-gallate (7CI), Epigallocatechol, 3-gallate, (-)- (8CI), Epigallocatechol, gallate (6CI), Gallic acid, 3-ester with epigallocatechol, (-)- (8CI), (-)-Epigallocatechin 3-O-gallate, (-)-Epigallocatechin 3-gallate, (-)-Epigallocatechin gallate, (-)-Epigallocatechol gallate, (-)-epi-Gallocatechin 3-O-gallate, 3-O-Galloyl-(-)-epigallocatechin, EGCG, Epigallocatechin 3-O-gallate, Epigallocatechin 3-gallate, Epigallocatechin gallate, Epigallocatechin-3-monogallate, L-Epigallocatechin gallate, NVP-XAA 723, PF-EGCg 90, Teavigo, Theliokeep, epi-Gallocatechin 3-O-gallate, epi-Gallocatechin gallate |
| InChI Formula | InChI=1S/C22H18O11/c23-10-5-12(24)11-7-18(33-22(31)9-3-15(27)20(30)16(28)4-9)21(32-17(11)6-10)8-1-13(25)19(29)14(26)2-8/h1-6,18,21,23-30H,7H2/t18-,21-/m1/s1 |
| SMILES | Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](Oc2c1)c4cc(O)c(O)c(O)c4 |
| IUPAC Name | [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate |
| Molecular Weight (g/mol) | 458.37 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?