Learn More
(-)-Epigallocatechin gallate, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J61745.MF
| Quantity | 50mg |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsEpigallocatechin gallate is reported to strongly inhibit Cu(2+)-mediated oxidative modification of LDLs. EGCG also inhibits NOS2 (iNOS).
Solubility
Soluble in ethanol, dimethyl formamide, water.
Notes
Specifications
| 989-51-5 | |
| MFCD00075940 | |
| 14,3526 | |
| Soluble in ethanol,dimethyl formamide,water. | |
| C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O | |
| 458.375 | |
| CHEBI:4806 | |
| (-)-Epigallocatechin gallate |
| C22H18O11 | |
| 50mg | |
| --epigallocatechin gallate, egcg, epigallocatechin gallate, epigallocatechin 3-gallate, tea catechin, epigallocatechin-3-gallate, --epigallocatechin-3-o-gallate, teavigo, --epigallocatechol gallate, epigallocatechin-3-monogallate | |
| WMBWREPUVVBILR-WIYYLYMNSA-N | |
| [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate | |
| 65064 | |
| 458.4 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.