missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Epigallocatechin Gallate, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
(-)-Epigallocatechin Gallate (EGCG) is a major polyphenol in green tea, which can inhibit cell proliferation and induce cell apoptosis. (-)-Epigallocatechin Gallate inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity. (-)-Epigallocatechin Gallate has a potent anticancer, antioxidant and anti-inflammatory properties against various types of cancers such as colorectal cancer, myeloid leukemia, thyroid carcinoma.
Specifications
Specifications
| Chemical Name or Material | (-)-Epigallocatechin Gallate |
| CAS | 989-51-5 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H18O11 |
| Quantity | 100 mg |
| Synonym | EGCG Epigallocatechol Gallate |
| Solubility Information | DMSO : ≥ 30 mg/mL (65.45 mM) ∣H2O : 20 mg/mL (43.63 mM; Need ultrasonic) |
| SMILES | O=C(O[C@H]1[C@@H](C2=CC(O)=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1)C4=CC(O)=C(O)C(O)=C4 |
| Molecular Weight (g/mol) | 458.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction