missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Epigallocatechin gallate, 95%

Product Code. 15224248 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15224248 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15224248 Supplier Thermo Scientific Chemicals Supplier No. 449010500
  • ONLINE EXCLUSIVE PRICE
    £84.10
    £75.65 / 50mg
    Save £8.45
    10% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 989-51-5 | C22H18O11 | 458.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 989-51-5
Color Pink to White
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C22H18O11
MDL Number MFCD00075940
Quantity 50 mg
Merck Index 153573
Synonym =--epigallocatechin gallate, egcg, epigallocatechin gallate, epigallocatechin 3-gallate, tea catechin, epigallocatechin-3-gallate, =--epigallocatechin-3-o-gallate, teavigo, =--epigallocatechol gallate, epigallocatechin-3-monogallate
Solubility Information Solubility in water: partially soluble. Other solubilities: practically insoluble in acetone,dmso,methanol
InChI Key WMBWREPUVVBILR-WIYYLYMNSA-N
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
IUPAC Name [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol) 458.37
PubChem CID 65064
ChEBI CHEBI:4806
Formula Weight 458.37
Percent Purity 95%
Physical Form Powder
Chemical Name or Material (-)-Epigallocatechin gallate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.