missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies (-)-Epigallocatechin Gallate

Product Code. 17170252
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 17170252

Brand: STEMCELL Technologies 73644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antioxidant and epigenetic modifier; Inhibits DNA methyltransferases (DNMTs)

  • (-)-Epigallocatechin Gallate is the most abundant polyphenol catechin antioxidant present in green tea (Frémont et al.; Johnson & Maddipati; Miller & Rice-Evans) and is known to inhibit DNA methyltransferases (DNMTs; IC50 = 0.21 - 0.47 μM; Lee et al.) (-)-Epigallocatechin Gallate also inhibits the formation of oxidized low-density lipoproteins (Yoshida et al.), which have a pathological role in cardiovascular diseases and atherosclerosis (Itabe et al.) (-)-Epigallocatechin Gallate has also been shown to inhibit peroxynitrite-mediated formation of 8-oxodeoxyguanosine and 3-nitrotyrosine (Fiala et al.)

Specifications

CAS 989-51-5
Synonym EGCG; NVP-XAA723; Tea catechin
Quantity 100 mg
Molecular Formula C22H18O11
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.