missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epigallocatechin-3-gallate 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | (-)-Epigallocatechin gallate |
| CAS | 989-51-5 |
| Analyte or Component Names | (-)-Epigallocatechin gallate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H18 O11 |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Benzoic acid, 3,4,5-trihydroxy-, (2R,3R)-3,4-dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-2H-1-benzopyran-3-yl ester, Benzoic acid, 3,4,5-trihydroxy-, 3,4-dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-2H-1-benzopyran-3-yl ester, (2R-cis)-, Epigallocatechol, 3-gallate (7CI), Epigallocatechol, 3-gallate, (-)- (8CI), Epigallocatechol, gallate (6CI), Gallic acid, 3-ester with epigallocatechol, (-)- (8CI), (-)-Epigallocatechin 3-O-gallate, (-)-Epigallocatechin 3-gallate, (-)-Epigallocatechin gallate, (-)-Epigallocatechol gallate, (-)-epi-Gallocatechin 3-O-gallate, 3-O-Galloyl-(-)-epigallocatechin, EGCG, Epigallocatechin 3-O-gallate, Epigallocatechin 3-gallate, Epigallocatechin gallate, Epigallocatechin-3-monogallate, L-Epigallocatechin gallate, NVP-XAA 723, PF-EGCg 90, Teavigo, Theliokeep, epi-Gallocatechin 3-O-gallate, epi-Gallocatechin gallate |
| InChI Formula | InChI=1S/C22H18O11/c23-10-5-12(24)11-7-18(33-22(31)9-3-15(27)20(30)16(28)4-9)21(32-17(11)6-10)8-1-13(25)19(29)14(26)2-8/h1-6,18,21,23-30H,7H2/t18-,21-/m1/s1 |
| SMILES | Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](Oc2c1)c4cc(O)c(O)c(O)c4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction