missing translation for 'onlineSavingsMsg'
Learn More

Epidermal Growth Factor Receptor/EGFR Ab-5 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.

£129.00 - £637.00

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-5
Clone H11
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Form Concentration Quantity Price Quantity & Availability  
Product Code Brand Form Concentration Quantity Price Quantity & Availability  
12640977
View Documents Promotion Details
Epredia
MS-316-P0
Concentrated 200μg/mL 100 μL
£129.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12650977
View Documents Promotion Details
Epredia
MS-316-P1
Concentrated 200μg/mL 500 μL
£354.00
500µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12630977
View Documents Promotion Details
Epredia
MS-316-P
Concentrated 200μg/mL 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12660977
View Documents Promotion Details
Epredia
MS-316-P1ABX
Concentrated 1mg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12603952
View Documents Promotion Details
Epredia
MS-316-PABX
Concentrated 1mg/mL 200 μL
£637.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: H11

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: HC2 20 d2 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells, Placenta or Squamous cell CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Epidermal Growth Factor Receptor/EGFR Ab-5
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Mouse
EGFR Family
HC2 20 d2 cells
Primary
H11
Monoclonal
Concentrated
RUO
Human
IgG1 κ
Videos
SDS
Documents

Documents

Certificates