missing translation for 'onlineSavingsMsg'
Learn More

Epidermal Growth Factor Receptor/EGFR Ab-15 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments.

£137.00 - £633.00

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-15
Clone H9B4
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12641967
View Documents
Epredia
MS-665-P0
100 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651967
View Documents
Epredia
MS-665-P1
500 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12631967
View Documents
Epredia
MS-665-P
1 mL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12671967
View Documents
Epredia
MS-665-P1ABX
100 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: H9B4

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Cytoplasmic domain

Immunogen: Purified EGFR from A431 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Epidermal Growth Factor Receptor/EGFR Ab-15
Immunoprecipitation
Unconjugated
Mouse
EGFR Family
Purified EGFR from A431 cells
Primary
H9B4
Monoclonal
Concentrated
RUO
Human
IgG1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.