missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EpiCult™-B Mouse Medium Kit

Product Code. 17118281
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17118281

Brand: STEMCELL Technologies 05610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit to culture and evaluate mouse mammary epithelial cells

  • EpiCult™-B (mouse) is a serum-free liquid culture medium optimized for the culture of mouse mammary luminal and myoepithelial cells
  • It is ideal for the culture and evaluation of mouse mammary epithelial progenitors in the mammary colony-forming unit assay when used in conjunction with an irradiated feeder layer such as NIH 3T3 cells
  • This medium is also used for enzymatic dissociation of mouse mammary tissue when supplemented with collagenase and hyaluronidase
  • Addition of Human Recombinant EGF (Catalog #78006), Human Recombinant bFGF (Catalog #78003), and Heparin Solution (Catalog #07980) is required for culturing cells

Specifications

Cell Type Mammary Cells
For Use With (Application) Cell Culture; Colony Assay
Quantity 1 Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.