missing translation for 'onlineSavingsMsg'
Learn More

Epichlorohydrin, TRC

Product Code. 30379488
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30379488

Brand: Toronto Research Chemicals TRCE58230050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Epichlorohydrin
CAS 106-89-8
Purity Grade Notes GC
Molecular Formula C3 H5 Cl O
Quantity 50 g
Synonym Oxirane, (chloromethyl)- (9CI), Propane, 1-chloro-2,3-epoxy- (6CI,8CI), 2-(Chloromethyl)oxirane, (Chloromethyl)ethylene oxide, (Chloromethyl)oxirane, (RS)-Epichlorhydrin, (±)-Epichlorohydrin, 1,2-Epoxy-3-chloropropane, 1-Chloro-2,3-epoxypropane, 10: PN: WO2017021334 SEQID: 16 claimed sequence, 2,3-Epoxypropyl chloride, 3-Chloro-1,2-epoxypropane, 3-Chloro-1,2-propylene oxide, 3-Chloropropene-1,2-oxide, 3-Chloropropylene oxide, Chloropropylene oxide, Epicerol, Epichlorohydrin, Epoxychloropropane, Glycerol epichlorohydrin, Glycidyl chloride, J 006, NSC 6747, dl-α-Epichlorohydrin, α-Epichlorohydrin, γ-Chloropropylene oxide
InChI Formula InChI=1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2
SMILES ClCC1CO1
IUPAC Name 2-(chloromethyl)oxirane
Molecular Weight (g/mol) 92.52
Formula Weight 92.0029
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.