missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin, MedChemExpress

Product Code. 16488946
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488946 100 mg 100mg
16464717 50 mg 50mg
16418818 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488946 Supplier Medchem Express Supplier No. HYN0001
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (-)-Epicatechin inhibits cyclooxygenase-1 (COX-1) with an IC50 of 3.2 μM. (-)-Epicatechin inhibits the IL-1β-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-κB.

(-)-Epicatechin inhibits cyclooxygenase-1 (COX-1) with an IC50 of 3.2 μM. (-)-Epicatechin inhibits the IL-1β-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-κB.

Specifications

Chemical Name or Material (-)-Epicatechin
CAS 490-46-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H14O6
Quantity 100 mg
Synonym (-)-Epicatechol Epicatechin epi-Catechin
Solubility Information DMSO : 100 mg/mL (344.51 mM; Need ultrasonic)
SMILES O[C@H]1[C@@H](C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1
Molecular Weight (g/mol) 290.27
Formula Weight 290.27
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.