missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin gallate, Thermo Scientific Chemicals

Product Code. 15463519
Change view
Click to view available options
Quantity:
10 mg
20 mg
Unit Size:
10mg
20mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463519 20 mg 20mg
15473519 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463519 Supplier Thermo Scientific Alfa Aesar Supplier No. J60814.F+
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A natural product from green tea that induces apoptosis

(-)-Epicatechin gallate is used as an antioxidant investigated as an adjunct treatment of some methyicillin resistant bacteria. Epicatechin gallate binds to the cell membrane in methicillin-resistant Staphylococcus aureus which decreases the fluidity of the bilayer and induces changes in gene expression. One of the catechin isomers and a potent antioxidant that can modulate a wide range of membrane proteins. Its bilayer-modifying potency was tested using gramicidin A (gA) channels as probes. All the catechins alter gA channel function and modify bilayer properties, with a 500-fold range in potency.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Epicatechin gallate is used as an antioxidant investigated as an adjunct treatment of some methyicillin resistant bacteria. Epicatechin gallate binds to the cell membrane in methicillin-resistant Staphylococcus aureus which decreases the fluidity of the bilayer and induces changes in gene expression. One of the catechin isomers and a potent antioxidant that can modulate a wide range of membrane proteins. Its bilayer-modifying potency was tested using gramicidin A (gA) channels as probes. All the catechins alter gA channel function and modify bilayer properties, with a 500-fold range in potency.

Solubility
Soluble in water, acetone, DMSO, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257-08-5
Molecular Formula C22H18O10
Molecular Weight (g/mol) 442.376
MDL Number MFCD00075936
InChI Key LSHVYAFMTMFKBA-TZIWHRDSSA-N
Synonym =--epicatechin gallate, epicatechin gallate, =--epicatechin-3-o-gallate, l-epicatechin gallate, =--epicatechin-3-gallate, =--epicatechingallate, epicatechin monogallate, --epicatechin 3-o-gallate, teatannin, epicatechol, gallate
PubChem CID 107905
ChEBI CHEBI:70255
IUPAC Name [(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O

Specifications

Color White
Quantity 20 mg
Solubility Information Soluble in water,acetone,DMSO,methanol.
Formula Weight 442.37
Physical Form Powder
Chemical Name or Material (-)-Epicatechin gallate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.