missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin gallate, MedChemExpress

Product Code. 16474717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474717 50 mg 50mg
16473826 25 mg 25mg
16428818 10 mg 10mg
16411268 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474717 Supplier Medchem Express Supplier No. HYN0002
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (-)-Epicatechin gallate (Epicatechin gallate) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 7.5 μM.

(-)-Epicatechin gallate (Epicatechin gallate) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 7.5 μM.

Specifications

Chemical Name or Material (-)-Epicatechin gallate
CAS 1257-08-5
Color White
Purity Grade Notes Research
Molecular Formula C22H18O10
Quantity 50 mg
Synonym Epicatechin gallate ECG (-)-Epicatechin 3-O-gallate
Solubility Information DMSO : ≥ 30 mg/mL (67.82 mM)
SMILES O=C(O[C@H]1[C@@H](C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1)C4=CC(O)=C(O)C(O)=C4
Molecular Weight (g/mol) 442.37
Formula Weight 442.37
Percent Purity 97.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.