missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin gallate, MedChemExpress

Product Code. 16428818
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16428818

Brand: Medchem Express HYN0002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-Epicatechin gallate (Epicatechin gallate) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 7.5 μM.

(-)-Epicatechin gallate (Epicatechin gallate) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 7.5 μM.

Specifications

Chemical Name or Material (-)-Epicatechin gallate
CAS 1257-08-5
Color White
Purity Grade Notes Research
Molecular Formula C22H18O10
Quantity 10 mg
Synonym Epicatechin gallate ECG (-)-Epicatechin 3-O-gallate
Solubility Information DMSO : ≥ 30 mg/mL (67.82 mM)
SMILES O=C(O[C@H]1[C@@H](C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1)C4=CC(O)=C(O)C(O)=C4
Molecular Weight (g/mol) 442.37
Formula Weight 442.37
Percent Purity 97.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.