missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin-3'-O-glucuronide, TRC

Product Code. 30387774
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387774 10 mg 10mg
30371964 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387774 Supplier Toronto Research Chemicals Supplier No. TRCE58233510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-Epicatechin-3'-O-glucuronide
CAS 603133-91-1
Purity Grade Notes HPLC
Molecular Formula C21 H22 O12
Quantity 10 mg
Synonym (-)-Epicatechin-3'-O-glucuronide, 5-[(2R,3R)-3,4-dihydro-3,5,7-trihydroxy-2H-1-benzopyran-2-yl]-2-hydroxyphenyl-β-D-glucopyranosiduronic acid, 5-[(2R,3R)-3,4-Dihydro-3,5,7-trihydroxy-2H-1-benzopyran-2-yl]-2-hydroxyphenyl β-D-glucopyranosiduronic acid, (-)-Epicatechin 3'-glucuronide, (-)-E
InChI Formula InChI=1S/C21H22O12/c22-8-4-11(24)9-6-12(25)18(31-13(9)5-8)7-1-2-10(23)14(3-7)32-21-17(28)15(26)16(27)19(33-21)20(29)30/h1-5,12,15-19,21-28H,6H2,(H,29,30)/t12-,15+,16+,17-,18-,19+,21-/m1/s1
SMILES O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O)c3
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[2-hydroxy-5-[(2R,3R)-3,5,7-trihydroxy-3,4-dihydro-2H-chromen-2-yl]phenoxy]oxane-2-carboxylic acid
Molecular Weight (g/mol) 466.392
Formula Weight 466.111
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.