missing translation for 'onlineSavingsMsg'
Learn More

Epibrassinolide, MedChemExpress

Product Code. 16479128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
16479128 10 mg 10mg
16459146 100 mg 100mg
16431538 5 mg 5mg
16424907 50 mg 50mg
16412396 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16479128 Leverantör Medchem Express Leverantörsnummer HYN0848

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Epibrassinolide (24-Epibrassinolide) is a ubiquitously occurring plant growth hormone which shows great potential to alleviate heavy metals and pesticide stress in plants. Epibrassinolide is a potential apoptotic inducer in various cancer cells without affecting the non-tumor cell growth.

Epibrassinolide (24-Epibrassinolide) is a ubiquitously occurring plant growth hormone which shows great potential to alleviate heavy metals and pesticide stress in plants. Epibrassinolide is a potential apoptotic inducer in various cancer cells without affecting the non-tumor cell growth.

Specifikationer

Chemical Name or Material Epibrassinolide
CAS 78821-43-9
Color White
Purity Grade Notes Research
Molecular Formula C28H48O6
Quantity 10 mg
Synonym 24-Epibrassinolide B1105 BP55
Solubility Information DMSO : 100 mg/mL (208.04 mM; Need ultrasonic)
SMILES C[C@@]1([C@@]2([H])[C@H](C)[C@@H](O)[C@H](O)[C@H](C)C(C)C)[C@](CC2)([H])[C@@](COC3=O)([H])[C@]([C@@]4([C@]3([H])C[C@H](O)[C@H](O)C4)C)([H])CC1
Molecular Weight (g/mol) 480.68
Formula Weight 480.68
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.