missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epiandrosterone ELISA Kit

Product Code. 17154853
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17154853

Brand: Invitrogen™ EIAANDO

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Epiandrosterone Competitive ELISA quantitates epiandrosterone in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.∣Principle of the method: The Epiandrosterone Competitive ELISA research-use-only kit is designed to quantitatively measure epiandrosterone independent of species. An epiandrosterone standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit epiandrosterone antibodies and epiandrosterone-peroxidase conjugate is added to the wells. The epiandrosterone-peroxidase conjugate and any epiandrosterone in the sample will compete to bind to the anti-rabbit epiandrosterone antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound epiandrosterone-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of epiandrosterone present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Epiandrosterone is a dehydroepiandrosterone metabolite and a precursor of testosterone and estradiol with hypolipidemic and anabolic property. Epiandrosterone, a potential neurosteroid, appears to bind to the gamma-aminobutyric acid (GABA)/benzodiazepine-receptor complex (GABA-RC), acting as a negative non-competitive modulator of GABA-RC as well as signal through the N-methyl-D-aspartate receptor. In addition this agent inhibits the pentose phosphate pathway (PPP) thereby dilating blood vessels pre-contracted by partial depolarization. Also, epiandrosterone inhibits the synthesis of thromboxane A2 in activated platelets, reduces plasma plasminogen activator inhibitor type 1 and tissue plasminogen activator antigen, increases serum levels of insulin-like growth factor 1 and increases cyclic guanosine monophosphate and nitric oxide synthesis. These effects may improve circulation in the microvasculature. [National Center for Biotechnology Information. PubChem Compound Database; CID=441302].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 3 beta-androsterone; 3 Beta-hydroxy-5 alpha-androstan-17-one; 5 alpha-androstan-3 beta-ol-17-one; C19H30O2; Epiandrosterone; Isoandrosterone; Trans-Androsterone
Assay Range 0.098 to 100 ng/mL
Assay Sensitivity 0.126 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 16%
Intraassay CV 8
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Storage Requirements -20°C
Target Epiandrosterone
Test Time 45 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.